Learn More
Avantor™ J.T. Baker™ NEUTRASORB™ Material for Acid Spills
Provides rapid, controlled neutralization of concentrated acids and visible color change as indication of complete neutralization
Brand: Avantor™ 4456-05
458.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : Weight : 4.00000kg
Description
- Caution: NEUTRASORB™ material should not be used for spills of hydrofluoric acid, fuming nitric acid, fuming sulfuric acid, or peroxy (per-) organic or other highly reactive acids.
- After complete neutralization, waste materials contain neutralized alkali metal and alkaline earth salts of the treated acid.
Specifications
Purple | |
Acid Spill Clean-up Kit | |
Bromocresol Purple Indicator Grade (CAS 115-40-2; < 0.1%), Calcium Carbonate (CAS 471-34-1; 5 - 25%), Magnesium Oxide (CAS 1309-48-4; 25 - 45%), Sodium Carbonate (CAS 497-19-8; 35 - 55%) |
Packed in box with foil Pk. | |
Granules | |
3.2 kg |