missing translation for 'onlineSavingsMsg'
Learn More

GPE Scientific™ J Young B40 Socket and High Vacuum Glass Key on the Outlet

GPE Scientific™ J Young B40 Socket and High Vacuum Glass Key on the Outlet is a basic vacuum line cold trap with a glass key on the outlet.

Brand:  GPE Scientific™ JYT/2

Product Code. 15804814

  • £491.00 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Solidly constructed for use as a basic vacuum line cold trap
  • Glass key on the outlet
Specifications

Specifications

Hose connection diameter: 15 mm
Body diameter: 44 mm
280 mm body
Inside tube diameter: 13 mm
Glass key on outlet
Basic vacuum line cold trap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers