Learn More
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint
GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand:  GPE Scientific™ GBF/28/12
 View more versions of this product
Product Code. 15834754
Description
Description
  • Spherical joint designed for use with ball joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Specifications
Shank Bore: 12 mm, Shank OD: 17 mm
Female Spherical Joint
28/12