Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
Brand:  GPE Scientific™ JC29
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15814784
  • £4.05 / Each
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Spring clip design
  • Securely fasten J Young spherical joints
Specifications
Specifications
Spring Clips for J Young Spherical Joints
28/12 and 28/15 spherical joints