Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
£138.00 - £167.00
Specifications
Includes | O-Ring Tap and O-Ring Ball Joint Cup |
---|---|
Product Type | Vacuum/Schlenk Flask |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15894734
|
GPE Scientific™
CSR/25 |
25 mL |
£138.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
15814744
|
GPE Scientific™
CSR/50 |
50 mL |
£139.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
15874734
|
GPE Scientific™
CSR/100 |
100 mL |
£139.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
15804744
|
GPE Scientific™
CSR/250 |
250 mL |
£144.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
15824744
|
GPE Scientific™
CSR/500 |
500 mL |
£150.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
15884734
|
GPE Scientific™
CSR/1000 |
1000 mL |
£167.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Round-bottomed shape
- Includes O-ring tap
- Includes O-ring ball joint cup
Spécification
O-Ring Tap and O-Ring Ball Joint Cup | |
Vacuum/Schlenk Flask |