missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Jack S1P Denim Shoe

Product Code. 17532586
Click to view available options
Size:
39
40
41
42
43
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17532586

Brand: Emma Safety Footwear MM91854941

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The unique heel design of the Jack Shoe is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.

  • S1P-Safetyshoe with sole plate for dry work
  • Metal Free
  • Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
  • The protective insole protects from sharp objects like nails and shards of metal.
  • This sneaker with fiberglass toe cap, is finished with robust denim. DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Manufacturing

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Blue
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Material Denim, Nubuck
Size 41
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.