Learn More
Hughes Jacketed and Insulated Tank Shower
Product Code. 17762348
Click to view available options
Capacity (Metric):
350 L
1500 L
Unit Size:
Each
Product Code. 17762348
Brand: Hughes HSHJ14K1SS
Only null left
Add to basket
Add to basket
Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates. Designed to meet and exceed ANSI standards by providing tepid water, even when there is no mains water supply.
  • Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates
  • Jacketed and insulated models maintain a steady temperature
  • Suitable for use where insufficient water pressure exists
  • Side mounted eye/face wash unit
  • Designed to meet and exceed ANSI standards by providing tepid water for over 15 minutes at a guaranteed flow of 76 liters per minute, even when there is no mains water supply
Specifications
Capacity (Metric) 1500 L
Material Stainless Steel
Product Type Tank Shower