missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Jackson Safety™ G40 Polyurethane Coated Gloves

Product Code. 15363342
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color Code Black
Color Black
Material Polyurethane
Product Type Gloves
Size X-Large
For Use With (Application) Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection
Quantity 60Pair
Resistant To Abrasion and tear
Series G40
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'