Learn More
Jagged1 Mouse anti-Human, Unconjugated, Clone: 2A7D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66890-1-IG-20UL
This item is not returnable.
View return policy
Description
The Jagged1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Jagged1. This antibody recognizes human antigen. The Jagged1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Jagged1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Flow Cytometry | |
2A7D11 | |
Unconjugated | |
20 μL | |
182 | |
IgA |