missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Janelia Fluor 549, free acid
Yellow dye (Reactive group: free acid). Applications: super resolution microscopy e.g. dSTORM & STED, confocal, flow and live cell imaging
Brand: Tocris Bioscience™ 6503/5
Additional Details : Weight : 0.00970kg
Description
Janelia Fluor® 549, free acid is a yellow fluorescent dye; supplied as a free acid. Used in protocol (2017 Grimm et al - see references below) for the synthesis of Janelia Fluor™ HaloTag™ and SNAP-Tag™ ligands, for use in live cell imaging experiments. Also suitable for flow cytometry. Excitation maximum = 549 nm; emission maximum = 571 nm; Quantum yield = 0.88; Extinction coefficient = 101,000 M-1cm-1.Specifications
3,6-Di-1-azetidinyl-9-(2,5-dicarboxyphenyl)xanthylium, inner salt trifluoroacetate | |
5 mg | |
C27H22N2O5.C2HF3O2 | |
90% |
2245946-45-4 | |
Free Acid Janelia Fluor™ Dyes | |
568.5 |