missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Janus Green B, pure, high purity biological stain

Product Code. 10163730
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2869-83-2
Molecular Formula C30H31ClN6
Molecular Weight (g/mol) 511.07
MDL Number MFCD00011758
InChI Key XXACTDWGHQXLGW-UHFFFAOYSA-M
Synonym C.I. 11050
PubChem CID 76123
ChEBI CHEBI:21184
IUPAC Name 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
SMILES [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C

Specifications

Dye Content 50% min.
Merck Index 15, 5303
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 511.07
Grade Pure
Packaging Glass bottle
Color Black or Brown/Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Janus Green B, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'