missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Japanese Encephalitis Virus Proteome Microarray

Japanese Encephalitis Virus Proteome Microarray with 3432 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.045.001

Additional Details : Weight : 0.02200kg

Product Code. 16367778

  • £482.30 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: Genome polyprotein cleaved into 13 chains: peptide 2k, capsid protein C, prM, peptide pr, small envelope protein M, envelope protein E, non-structural protein 1, non-structural protein 2A, serine protease subunit NS2B, serine protease NS3, non-structural protein 4A, non-structural protein 4B, RNA-directed RNA polymerase NS5
  • Peptide Length/Overlap: 15 aa/ 14 aa
  • Number of Peptides: 3432 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers