Learn More
Fritsch™ Jaw Crusher Pulverisette 1
Brand:  Fritsch™ 01.5020.00
Code : 04
Additional Details : Weight : 177.00000kg
Product Code. 13114704
  • POA / Each
Estimated Shipment: 04-10-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Model 1 jaw crusher feed particle size approx 60mm, throughput 140kg/h Gap width(fineness)1-15mm, 41x83x72cm(wxdxh) 65mm x 65mm funnel opening 230V 50/60Hz 1570W 3 phase Fritsch