missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ Conductivity Standards
Click to view available options
Quantity:
500 mL
Conductivity:
12.88 mS
84 μS
Description
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
Specifications
Specifications
Conductivity | 12.88 mS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction