Learn More
Cole-Parmer™ Jenway™ Cuvette Holders
Product Code. 13246389
Click to view available options
For Use With (Equipment):
63/73/ 67 Series Spec
6850 Series Spectrophotometers
Pathlength:
10 to 100 mm
10 x 10 mm
Unit Size:
Each
Product Code. 13246389
Brand: Cole-Parmer™ 8307053