Learn More
Cole-Parmer™ Jenway™ Cuvette Holders
Brand:  Cole-Parmer™ 83070-57
Code : A0
Additional Details : Weight : 0.39600kg
 View more versions of this product
Product Code. 13266389
  • £409.91 / Each
Estimated Shipment: 06-08-2024
to see stock.
Specifications
Cuvette Holder
6850 Series Spectrophotometers
10 to 100 mm