Learn More
Cole-Parmer™ Jenway™ Glass Flow-through Cell
Brand:  Cole-Parmer™ 83070-50
Product Code. 12018856
  • £700.00 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Flow-through Cell
80 μL
Glass
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Glass Flow-through Cell > Capacity: 80μL