Learn More
Jenway™ Model 731001 Visible Single Beam Spectrophotometer
Ideal for routine testing in clinical, veterinary, pharmaceutical and QC laboratories.
Brand:  Jenway™ 83058-17
3301.25 GBP valid until 2025-12-31
Use promo code "24356" to get your promotional price.
Product Code. 13101744
  • £3475.00 / Each
Explore more special offers
Description
Description
The 73 series spectrophotometers use icon-driven software and have an improved navigation system for easy and intuitive usability. The 73 series range is comprised of four models: Models 7300 and 7305 are basic non-scanning spectrophotometers with measurement modes for absorbance, % transmittance and concentration. This model 7310 and model 7315 are advanced scanning spectrophotometers which have additional measurement modes for spectrum scanning, kinetics and quantitation. The 7310 and 7315 have the additional feature of an easy-access USB port which enables results and methods to be saved directly to USB memory stick.
  • Icon-driven software
  • Small footprint
  • Autologging capabilities
  • Save methods and results to USB memory stick
  • Three year warranty
  • Extensive range of accessories available The 73 series spectrophotometers are ideal for routine testing in clinical, veterinary, pharmaceutical and QC laboratories.
Specifications
Specifications
–300 to 9999
Tungsten Halogen
Visible Spectrophotometer
220 mm
1 nm
±2 nm
275 mm
400 mm
5 nm
320 to 1000 nm
Single Beam
3 Years
6 kg
Product Suggestions
Product Suggestions
Videos
Videos
  • Jenway Introducing the 73 series spectrophotometers
  • Jenway Operation of a 73 series spectrophotometer
  • Jenway Adding a printer to a 73 series spectrophotometer
  • Jenway 73 spectrophotometer accessories
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Jenway™ Model 731001 Visible Single Beam Spectrophotometer > Model: 731001