Learn More
Bibby Scientific™ Jenway™ Paper Roll
Replace paper in Jenway™ Bench pH Meters and 73 Series Spectrophotemeter printers quickly and easily. Bibby Scientific™ Jenway™ Paper Roll saves waste and ink because it uses heat, not ink to print.
Brand:  Bibby Scientific™ 99959-81
Code : 03
Additional Details : Weight : 0.02340kg
Product Code. 11420479
  • £10.32 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • For use with Jenway™ Bench pH Meters, 73 Series Spectrophotemeter printers
  • Thermal paper prints with heat, not ink.
Specifications
Specifications
Jenway™ Bench pH meters, 73 series Spectrophotometers, Dissolved Oxygen Bench Meters and other Jenway™ printers.
Paper Roll
1 Ea.