missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ pH Temperature Electrode
Jenway™ pH Temperature Electrodes are used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
£96.51 - £204.00
Specifications
Connector Types | BNC |
---|---|
Diameter (Metric) | 12 mm |
Dimensions (L x W) | 12 dia. x 120 mm L |
Type | Combination |
Length (Metric) | 120 mm |
Product Code | Brand | For Use With (Application) | Material | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Material | Description | Price | Quantity & Availability | ||||
11420499
|
Jenway™
99960-43 |
Measurements of food extracts and agar plates, flat head makes it ideal for surface measurements | Epoxy | Flat end type |
£204.00
Each |
In Stock
Log in to see stock. |
||||
11440479
|
Jenway™
99960-35 |
General purpose pH measurement for accurate Q.C. and research work | Glass | Bulb end type |
£96.51
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
- Used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
- Combination Type
- Refillable
- Reference Type: AgCl
- Isopotential: 0±20mV
Specifications
BNC | |
12 dia. x 120 mm L | |
120 mm | |
0°C to 80°C |
12 mm | |
Combination | |
0 to 14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Jenway™ pH Temperature Electrode