man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Lab Electrical Equipment
Multi-Purpose Power Supplies
Jenway™ Power Supply
Print
Jenway™ Power Supply
£28.61
See promotional offers below
Products
1
Description
Videos
SDS
Documents
Product Certifications
Products
1
Product Code
Brand
For Use With (Equipment)
Price
Quantity & Availability
Product Code
Brand
For Use With (Equipment)
Price
Quantity & Availability
12943247
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Jenway™
99956-00
pH meters
£28.61
Each
View alternative products
Save Up to
»
In Stock
Log in
to see stock.
Add to basket
Description
Description
Plug type: UK plug
Videos
SDS
Documents
Documents
Product Certifications
Back to Top