missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jerrican Blue HDPE 30L Stackable 61mm Tamper-Evident Neck
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Click to view available options
Material:
HDPE
Description
UN Certified container manufactured from high density polyethylene (HDPE). Ideal for storing, transporting and decanting liquids including hazardous products, offering maximum pallet utilisation.
Specifications
Product Type | Evident Neck |
Color | Blue |
For Use With (Application) | Storing, Transporting and Decanting Liquids |
Material | HDPE |
Weight (Metric) | 10 kg |
Certifications/Compliance | UN Certified Container Manufactured from High Density Polyethylene (HDPE) |
Shape | Rectangular |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'