Learn More
Invitrogen™ JetFlex™ Genomic DNA Purification Kit
The JetFlex™ Genomic DNA Purification Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), mammalian cells, tissues, plant tissues, bacteria, and yeast.
Brand: Invitrogen™ A30700
186.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Single-tube technology allows for isolation of high quality, high molecular weight, RNA-free gDNA without the use of spin columns of a vacuum manifold.
Key benefits include:
- Large volume processing of 2 mL of whole blood or other body fluid
- Capable of processing:
- 2 x 107 mammalian cells
- 10 mg of tissue
- 500 μL bacterial culture
- 2 x 108 cells/mL yeast culture
- Simple protocol that does not require columns or vacuum manifolds
Specifications
• 1 x 33 mL RBC Lysis Buffer, Store at Room Temperature • 1 x 33 mL Cell Lysis Buffer, Store at Room Temperature • 1 x 17 mL PPT Buffer, Store at Room Temperature • 1 x 8 mL Pellet Compactor, Store at Room Temperature • 1 x 22 mL TE Buffer, Store (at Room Temperature • 2 x 21 mg Proteinase K (lyophilized powder), Store 2°C to 8°C • 1 x 1.1 mL RNase A, Store 2°C to 8°C |
|
Tube Centrifugation | |
Real-Time PCR, PCR | |
100 Preps | |
Genomic DNA Purification Kit | |
Room Temperature |
Spin Column | |
Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum) | |
Not High-throughput Compatible | |
JetFlex™ | |
100 Preps | |
Genomic DNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.