missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JetFlex™ Genomic DNA Purification Kit
Click to view available options
No. of Reactions:
100 Preps
500 Preps
Unit Size:
Pack of 100
Pack of 500
Beskrivning
Single-tube technology allows for isolation of high quality, high molecular weight, RNA-free gDNA without the use of spin columns of a vacuum manifold.
Key benefits include:
- Large volume processing of 2 mL of whole blood or other body fluid
- Capable of processing:
- 2 x 107 mammalian cells
- 10 mg of tissue
- 500 μL bacterial culture
- 2 x 108 cells/mL yeast culture
- Simple protocol that does not require columns or vacuum manifolds
Specifikationer
Specifikationer
| Content And Storage | • 1 x 165 mL RBC Lysis Buffer, Store at Room Temperature • 1 x 165 mL Cell Lysis Buffer, Store at Room Temperature • 1 x 83 mL PPT Buffer, Store at Room Temperature • 1 x 30 mL Pellet Compactor, Store at Room Temperature • 1 x 110 mL TE Buffer, Store (at Room Temperature • 10 x 21 mg Proteinase K (lyophilized powder), Store 2°C to 8°C • 5 x 1.1 mL RNase A, Store 2°C to 8°C |
| Format | Spin Column |
| Isolation Technology | Tube Centrifugation |
| Sample Type | Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum) |
| For Use With (Application) | Real-Time PCR, PCR |
| High-throughput Compatibility | Not High-throughput Compatible |
| No. of Reactions | 500 Preps |
| Product Line | JetFlex |
| Product Type | Genomic DNA Purification Kit |
| Quantity | 500 Preps |
| Visa mer |
For Research Use Only. Not for use in diagnostic procedures.
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering