Learn More
Invitrogen™ JetQuick™ Blood and Cell Culture DNA Midiprep Kit
The JetQuick™ Blood and Cell Culture DNA Midiprep Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), and mammalian cells.
£296.00 - £472.00
Specifications
Format | Kit |
---|---|
Isolation Technology | Spin Column |
Scale | Midi |
Sample Type | Blood, Cells |
For Use With (Application) | Real-Time PCR, PCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | ||||||
15667878
|
Invitrogen™
A30704 |
50 Preps | N/A | N/A | N/A | |||||
15657878
|
Invitrogen™
A30703 |
20 Preps |
£296.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Utilizes silica membrane-based technology that allows for isolation of 20-50 kb DNA that is suitable for a wide range of downstream applications, including PCR, restriction enzyme digestion, and Southern blotting.
Key benefits include:
• Large volume processing, up to 3 mL of blood or saliva
• Long double-stranded DNA (20-50 kb) recovery
• Compatible with centrifugation or a vacuum manifold
The midiprep format enables the processing of up to 3 mL of whole blood or other body fluid. DNA from up to 1 x 108 mammalian cells can be isolated. The purified DNA is ready for downstream analysis.
Specifications
Kit | |
Midi | |
Real-Time PCR, PCR | |
JetQuick™ | |
Room Temperature |
Spin Column | |
Blood, Cells | |
Not High-throughput Compatible | |
Blood and Cell Culture DNA Kit | |
Genomic DNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.