Learn More
Cole-Parmer™ Jiffy-Jack Lab Jack
Jiffy-Jack Lab Jack features excellent side-to-side rigidity and support.
Brand:  Cole-Parmer™ 08057-20
237.62 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 11719128
  • £258.32 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Jiffy-Jack Lab Jack features excellent side-to-side rigidity and support.
  • Provide stable, height-adjustable support for hot plates, baths, glassware, and many other items
  • Positioners are constructed of rugged, chemical-resistant anodized aluminum with stainless steel operating screw.
  • Top-plate skid pad prevents slipping.
  • Each positioner is precision engineered for smooth, continuous operation throughout its range.
  • Easily set the exact height needed with the turn of a knob.
  • A 10 mm tapped hole in the right rear corner accommodates a frame rod (not included) for mounting ring stands and other lab apparatus
  • Maximum support static load is 132 lb. (60 kg)
Specifications
Specifications
Aluminum (Positioner), Stainless Steel (Operating Screw)
131.9 lb.
1
6 x 6 in. (Top)
132 lb. capacity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
Cole-Parmer™ Jiffy-Jack Lab Jack > lab jack