missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526399
This item is not returnable.
View return policy
Description
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt]Specifications
JMJD2B | |
Polyclonal | |
Unconjugated | |
KDM4B | |
4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O94953 | |
KDM4B | |
KLH conjugated synthetic peptide between 4-31 amino acids from the N-terminal region of human JMJD2B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |