missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNJ 10397049, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective OX2 receptor antagonist (pIC50 = 7.4 for chimeric OX2 receptors; pKB values are 5.9 and 8.5 for OX1 and OX2 receptors respectively). Shows no significant activity in a panel of over 50 other neurotransmitters and neuropeptide receptors. Achieves high level of OX2 receptor occupancy in the rat brain; exhibits sleep-promoting effects in rats.
Specifications
Quantity | 50 mg |
Formula Weight | 484.18 |
Percent Purity | >99% |
Chemical Name or Material | JNJ 10397049 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'