missing translation for 'onlineSavingsMsg'
Learn More

JNJ 5207852 dihydrochloride, Tocris Bioscience™

Product Code. 18035222
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity H3 receptor antagonist

High affinity histamine H3 receptor neutral antagonist (pKi values are 8.9 and 9.2 in rat and human respectively). Brain penetrant and orally active. Has 3- and 100-fold higher affinity than thioperamide (Cat. No. 0644) for rat and human H3 receptors respectively. Suppresses slow-wave sleep; exhibits wake-promoting effects in rodent arousal models.

Chemical Identifiers

CAS 398473-34-2
Molecular Formula C20H32N2O
Molecular Weight (g/mol) 316.489
InChI Key PTKHFRNHJULJKT-UHFFFAOYSA-N
Synonym unii-4i9ovb1g7d, 4i9ovb1g7d, 1-4-3-piperidinopropoxy benzyl piperidine, 1-4-3-piperidin-1-yl propoxy benzyl piperidine, 1-3-4-piperidin-1-ylmethyl phenoxy propyl piperidine, 1-3-4-piperidin-1-yl methyl phenoxy propyl piperidine, piperidine, 1-3-4-1-piperidinylmethyl phenoxy propyl, jnj 5207852 dihydrochloride
PubChem CID 2766326
IUPAC Name 1-[3-[4-(piperidin-1-ylmethyl)phenoxy]propyl]piperidine
SMILES C1CCN(CC1)CCCOC2=CC=C(C=C2)CN3CCCCC3

Specifications

Quantity 10 mg
Formula Weight 389.4
Percent Purity >99%
Chemical Name or Material JNJ 5207852 dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'