missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNJ 5207852 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity histamine H3 receptor neutral antagonist (pKi values are 8.9 and 9.2 in rat and human respectively). Brain penetrant and orally active. Has 3- and 100-fold higher affinity than thioperamide (Cat. No. 0644) for rat and human H3 receptors respectively. Suppresses slow-wave sleep; exhibits wake-promoting effects in rodent arousal models.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 389.4 |
Percent Purity | >99% |
Chemical Name or Material | JNJ 5207852 dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'