Learn More
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette
BIOPHARMA
Quick and easy to use. It is ideal for taking samples from low viscosity liquids.
£192.00 - £282.00
Products 3
Product Code Brand Capacity (Metric) Length (Metric) Sterility Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Sterility Price Quantity & Availability  
11970008
BIOPHARMA
View Documents
Sampling Systems™
8051H-450
80 mL 450 mm Non-sterile
£192.00
Pack of 50
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
11900018
BIOPHARMA
View Documents
Sampling Systems™
8051H-900
180 mL 900 mm Non-sterile
£252.00
Pack of 50
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
11910018
BIOPHARMA
View Documents
Sampling Systems™
8051H-900S
180 mL 900 mm Sterile
£282.00
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
Description
Description
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas (450mm and 900mm lengths only)
  • Available in 3 different sizes
  • Highly visible color coded handle
Operation:
  • Press thumb onto blue end of the tube until lowered to the correct sampling depth.
  • At the required depth raise thumb until levels equalize.
  • Press the thumb onto tube to seal in the sample.
  • Remove from the product.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.