Learn More
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
Brand:  Trajan™ 009920
 View more versions of this product
Product Code. 10751042
  • £684.32 / Each
Explore more special offers
Description
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Specifications
Acrylic, Acrylic (Barrel)
For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Jumbo Syringe
1000 mL
100 mm
159.1 mm
1000MAR-LL-GT
Trajan
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan™ Jumbo Syringe > Capacity: 1L