missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Jumbosep™ Centrifugal Device Starter Kits

Jumbosep™ Centrifugal Device Starter Kit replaces dialysis, chemical precipitation, and lyophilization.

Brand:  Cytiva FD000K65

 View more versions of this product

Product Code. 17124721

  • £185.00 / Pack of 4
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
  • Provides high recoveries, typically > 90%
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EFA: 15.2 cm2
4/Pk.
20 to 60 mL (Sample)
Non-sterile
Generic Starter Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Jumbosep™ Centrifugal Device Starter Kits > Generic Starter Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.