missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunB Recombinant Rabbit Monoclonal Antibody (15H19L4), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701702
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Monkey, Rat, Mouse and Feline. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.Specifications
JunB | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P17275 | |
Junb | |
Peptides corresponding to Human JUNB (aa 6-25, 262-279) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Immunocytochemistry, Western Blot | |
15H19L4 | |
Unconjugated | |
Junb | |
activator protein 1; AP-1; Jun B; jun B proto-oncogene; Junb; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; myD21; Oncogene Jun-B; Transcription factor jun-B | |
Rabbit | |
Protein A | |
RUO | |
3726 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |