Learn More
3M™ Jupiter™ Starter Kits
Ready-to-use starter kits feature everything needed for filtration in the laboratory. Complete with electronic audible and visual alarms in the event of low battery and/or low air flow.
£962.50 - £1372.60
Specifications
Battery Life | 8 hours |
---|---|
Battery Type | NiMH |
Filter Type | OV/HE Filter |
Product Type | PAPR Assembly, No Headtop |
For Use With (Application) | Protection against gases, vapors, and particulates, or a combination of contaminants per filter used |
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
11741403
|
3M™
7000079735 |
OV/HE Filter | N/A | ||||||
13443696
|
3M™
7000044295 |
OV/HE Filter | N/A | ||||||
Description
- High comfort decontamination belt
- Air Flow Indicator and Calibration tube
- Pair of Organic Vapour and Particulate (A2P) Filters and Prefilters
- Length-adjusting breathing tube
- 8-hour rechargeable battery (NiMH) and a battery charger
Specifications
8 hours | |
OV/HE Filter | |
Protection against gases, vapors, and particulates, or a combination of contaminants per filter used |
NiMH | |
PAPR Assembly, No Headtop | |
1 Ea. |