missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Bottle Jacket

Provides added protection of a polyethylene bottle without contaminating high-purity solvents

Brand:  Justrite™ 12009

Product Code. 13386309

  • £43.45 / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for 4L and 1 gal. bottles
  • Place a bottle in the bottom half of the jacket and rotate lid for secure containment
  • Handle offers balanced pouring
  • Acts as a shock absorber to minimize breakage and spills due to bumps or falls

Specifications

Specifications

Bottle Jacket
Bottles (4 L, 1 gal.)
35.5 cm
14 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.