Learn More
Justrite™ Brass Faucet
Self-closing safety faucets prevents flashback of fire into drum
Brand:  Justrite™ 08540
Product Code. 10478022
  • £69.70 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features polytetrafluoroethylene (PTFE) seal
  • Adjustable models screw tightly into drum
  • Faucet extensions provide extra dispensing accuracy and prevents spills
Certifications
FM Global tested and approved
Specifications
Specifications
FM
Self Close
0.75 in.
Brass Control Flow Lab Safety Faucet
Brass
1.90 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products