missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ EcoPolyBlend™ Spill Control Pallets

Made of up to 100% recycled polyethylene

Brand:  Justrite™ 28635

 View more versions of this product

Product Code. 11939385

  • £499.00 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Two and four drum square pallets include wide forklift pockets.
This item is not returnable. View return policy

Description

Description

  • Protect against groundwater contamination, using post-industrial material
  • Robust pallets withstand the toughest environments for superior spill control
  • Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
  • Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
  • Oversized leakproof sumps meet or exceed regulations
  • Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
  • Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
  • Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
  • Square pallet may be equipped with an optional ramp
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail

Compliance

Meet or exceed EPA 40 CFR 264.175; NFPA 1 the Uniform Fire Code 60.3.2.10; International Fire Code 2704.2.3; Spill Prevention, Control and Countermeasures (SPCC) Compliant; NFPA 30 9.13.1 and OSHA 29 CFR 1910.106(e)(2)(iii).

Specifications

Specifications

SPCC, EPA, NFPA, U (NFPA), meets fire codes
73 gal.
Polyethylene
Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination
4 Drums
5000 lb.
Chemical
124.5 cm
26 cm
124.5 cm
Black
276 L
Ecopolyblend Spill Control Pallet
Drums
4-Drum Square
2268 kg
49 in.
10.25 in.
49 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.