Learn More
Invitrogen™ K-MetStat Panel, SNAP-ChIP™ Spike-in
SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation), a proprietary technology from EpiCypher, uses DNA-barcoded semi-syntheic nucleosomes bearing distinct post-translational as spike-in ChIP controls.
£395.00
Specifications
Content And Storage | Store at -5°C to -30°C. |
---|---|
Description | Recombinant Nucleosome Panel |
Product Line | K-MetStat™, SNAP-ChIP™ |
Product Type | SNAP-ChIP Panel |
Description
SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation), a proprietary technology from EpiCypher, uses DNA-barcoded semi-syntheic nucleosomes bearing distinct post-translational as spike-in ChIP controls.
- Homogenous, fully defined standards faithfully represent target mononucleosomes in the experimental sample
- Nucleosomes are subjected to rigorous quality control for lot-to-lot consistency
- Unique DNA barcodes can be distinguished from experimental sample genomes
- Spike-ins provide a direct readout of antibody performance in ChIP
- Ability to monitor technical variability between samples
- Defined standards enable universal normalization across experiments
SNAP-ChIP panels are directly compatible with your current ChIP workflow, with semi-synthetic nucleosomes bearing the post-translational modification of interest immunoprecipitated and processed alongside sample chromatin.
Users can directly monitor antibody capability (specificity and enrichment) in a ChIP experiment by measuring the recovery of on- and off-target nucleosomes. The SNAP spike-ins can be further used as a defined standard to normalize your ChIP data, thereby controlling for technical variability across experiments.
The K-MetStat Panel is a lysine methylation status panel that consists of a pool of nucleosomes carrying fifteen well-studied, disease relevant lysine methyl marks on histones H3 and H4 (mono-, di- and tri-methylated H3K4, H3K9, H3K27, H3K36, and H4K20) plus an unmodified control. Further, each of these nucleosomes is wrapped by two independent DNA barcodes (32 total barcodes in the panel), allowing for an internal technical replicate in each ChIP reaction. A single spike-in of the panel allows users to check antibody specificity by examining the post-IP recovery of on- versus off-target SNAP-ChIP nucleosomes, supporting the generation of high quality ChIP data.
Specifications
Store at -5°C to -30°C. | |
Recombinant Nucleosome Panel | |
K-MetStat™, SNAP-ChIP™ | |
SNAP-ChIP Panel |
For Research Use Only. Not for use in diagnostic procedures.