Learn More
Hanna Instruments™ K-Type Waterproof Thermocouple Thermometers
A digital, waterproof thermometer that offers high accuracy temperature measurements using a wide variety of interchangeable probes.
Marke: Hanna Instruments™ HI935005
Code : A0
Additional Details : Weight : 0.47700kg
Description
- K-Type Thermocouple Thermometer is a digital, waterproof thermometer that can be used with a wide variety of K-type probes
- Offers high accuracy temperature measurements using a wide variety of interchangeable probes
- Offers two measurement ranges from -50.0 to 199.9°C and 200 to 1350°C which can also be displayed in °F (-58.0 to 399.9°F and 400 to 2462°F)
- With a ±0.2% full scale accuracy, is perfectly suited for temperature measurements in the laboratory or the field
Specifications
Yes |