missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90
Shop All asecos Products
Click to view available options
Model:
K90.060.110.050.UB.2T
K90.060.110.050.UB.ST
K90.060.140.050.UB.3T
K90.060.140.050.UB.ST
Weight (Metric):
152.50 kg
186.50 kg
190.50 kg
196.50 kg
No. of Doors:
1
2
3
No. of Shelves:
1
2
Unit Size:
Each
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
- Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
- No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
- Certificates
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- CE compliant
Specifications
Specifications
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
| Configuration | 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), 1 x pull-out shelf (panel melamine resin-coated/PP) |
| Depth (Metric) Exterior | 502 mm |
| Depth (Metric) Interior | 380 mm |
| Dimensions (D x W x H) Interior | Left: 380 x 770 x 502 mm, Right: 468 x 440 x 562 mm |
| Dimensions (D x W x H) | 502 x 1402 x 601 mm |
| Door Style | Wing |
| For Use With (Application) | Safe and approved storage of aggressive and flammable hazardous substances in working areas |
| Height (Metric) Exterior | 601 mm |
| Height (Metric) Interior | 502 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction