missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ KA1/GRIK4/Glutamate Receptor KA1 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Brand:  Novus Biologicals™ NBP2-62686PEP

Additional Details : Weight : 0.00970kg

Product Code. 18210173

  • £178.18 / 100µL
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GRIK4. The KA1/GRIK4/Glutamate Receptor KA1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

2900
>80% by SDS-PAGE and Coomassie blue staining
PBS and 1M Urea, pH 7.4.
GRIK4
100 μl
E. coli
Human
Store at -20°C. Avoid freeze-thaw cycles.
Antibody Competition
21 kDa
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54474. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers