Learn More
WTW™ Kal.Kit Calibration Set for Turb™ 555T Turbidity Meter
Dedicated Calibration Set for the WTW™ Turb™ 555T Turbidity Meter includes indexing rings and cleaning tissues
Brand: WTW™ 600545
This item is not returnable.
View return policy
Description
- Calibration set comprising four vials at 0.02, 10.0, 100 and 1750 NTU
- For use with the Turb™ 555 benchtop advanced laboratory turbidimeter
- Kit includes tissues and indexing rings
Specifications
Turbidity Standard | |
Turb 555 Turbidity Meter | |
Four vials at 0.02, 10.0, 100 and 1750 NTU, cleaning tissues and indexing rings |