missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Pure, Light, Washed with Acid, Fisher Chemical™

Product Code. 10753991
Click to view available options
Quantity:
3 kg
Packaging:
Plastic keg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Melting Point 1750°C
Color Beige
Physical Form Powder/Solid
Quantity 3 kg
MDL Number 62311
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
Formula Weight 258.2g/mol
Packaging Plastic keg
Product Identifier
  • Kaolin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'