missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15202-1-AP-150UL
This item is not returnable.
View return policy
Description
TIF1 beta is a member of the tripartite motif (TRIM) family. It is composed of a N-terminal RBCC (RING finger, B boxes, coiled coil) motif, and a C-terminal bromodomain which is preceded by a PHD finger. TIF1 beta has been reported to function as a co-repressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins. As such, it plays a crucial role in early embryogenesis. It appears that the co-repressor function of TIF1 beta is related to its organization and/or maintece of higher-order, heterochromatin-like structures.Specifications
KAP1 | |
Polyclonal | |
Unconjugated | |
TRIM28 | |
KAP 1, KAP1, KRAB associated protein 1, KRAB interacting protein 1, KRIP 1, Nuclear corepressor KAP 1, RING finger protein 96, RNF96, TF1B, TIF1 beta, TIF1B, TRIM28, tripartite motif containing 28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10155, 116698, 21849 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08629, Q13263, Q62318 | |
TRIM28 | |
KAP1 Fusion Protein Ag7350 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |