missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
£133.00 - £205.00
Specifications
Color | Colorless |
---|---|
Packaging | Amber glass bottle |
Physical Form | Liquid |
Vapor Density | > 1.0 |
MDL Number | 81734 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11443813
|
Fisher Chemical
K/2300R/08 |
500 mL | Amber glass bottle |
£133.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205592
|
Fisher Chemical
K/2300R/15 |
1 L | Amber glass bottle |
£205.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Matrix to be used in conjunction with Complete 5K (US: AL2250R; EU: K/2250R) for high water content analysis of aldehydes and ketones.
Specifications
Colorless | |
Liquid | |
81734 |
Amber glass bottle | |
> 1.0 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Aqualine Matrix K
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Chloroform; 2-Butoxyethanol; Ethyl alcohol; Ethylene glycol
- REA17-CMR1-Restricted to professional users