missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Reagent Solution A, for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol), Honeywell™ Fluka™

for pyridine-based volumetric Karl Fischer titration (pyridine, sulfur dioxide, methanol)

Brand:  Honeywell Fluka 36116-1L

Code : NEW

Additional Details : Weight : 1.01000kg Transport : UN number : 1992 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15695110

  • £60.31 / 1 L
Estimated Shipment: 14-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
Specifications

Specifications

16°C (60.8°F)
Karl Fischer reagent solution A
Glass bottle
MFCD00004595
CO
32.04
1 L
1.01g/mL
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Karl Fischer reagent solution A

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, pyridine,sulphur dioxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers