missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ KaryoMAX™ Colcemid™ Solution in PBS

KaryoMAX™ Colcemid™ Solution in PBS

Brand:  Gibco™ 15212012

Product Code. 10116784

  • £33.14 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is a 10μg/ml N-desacetyl-N-methylocolchicine (Colcemid™) solution made in Phosphate Buffered Saline (PBS). It is used for arresting cells in metaphase so the chromosomes can be separated for cytogenetic studies and in vitro diagnostic procedures. Colcemid™ prevents spindle formation during mitosis, causing metaphase arrest. The mechanism of action is similar to that of colchicine, but with lower mammalian toxicity. The complete formulation is available.

Product Use
For in vitro diagnostic use.

cGMP Manufacturing and Quality System
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Liquid
Sterile-filtered
6 to 8
10 mL
Mammalian Cells
Chromosome Analysis Media
Clinical Research, Translational Research
KaryoMAX™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.