Learn More
KaryoStat™ Assay
Shop All Applied Biosystems ProductsDescription
The assay enables the detection of aneuploidies, submicroscopic aberrations, and mosaic events for research use. Because the assay provides genotype information, it can help in the identification of triploidies and copy neutral events such as absence of heterozygosity. Our proprietary manufacturing technology produces arrays that are highly reproducible between each batch with no risk of probe dropout inherent in bead manufacturing techniques. The assay includes arrays, reagents, and easy-to-use data analysis software for a cost-effective and streamlined analysis of pluripotent stem cell lines.
Features include:
- Cost effective and accurate detection of chromosomal abnormalities
- Karyotyping and genotyping (sample ID) with one assay
- Simple analysis; no cytogenetics expertise necessary
- Assay includes arrays, reagents, and free downloadable software
- 2.5 days to results
Specifications
Specifications
| Type | Assay |
| Number of Arrays | 24 arrays |
| Format | Genechip Probe Array |
| Species | Human |
| Content And Storage | Store at 2–8°C: • Arrays • Module 2: Pre-Lab TE Buffer and Water • Module 4: Main Lab Stain, Holding Buffer, Beads, and Water Store at -5 to -30°C: • Module 1: Pre-Lab Restriction, Ligation, and Amplification • Module 3: Main Lab Fragmentation, Labeling, and Hybridization Store at room temperature: • Module 5: Main Lab Elution Buffer • Purification Wash Buffer • Wash Buffer A • Wash Buffer B |
| Includes | Arrays, Reagents, Data Analysis Software |
| Product Line | KaryoStat |
| Quantity | 24 assays |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.