missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ KBM™- 2 Keratinocyte Basal Medium-2
Optimized for the proliferation of keratinocytes in a serum-free environment and typically provides higher growth rates than KGM™ Medium.
Brand: Lonza™ CC-3103
Additional Details : Weight : 1.00000kg
Description
- Basal Medium - contains no growth factors cytokines, or supplements.
- Optimized for the proliferation of keratinocytes in a serum-free environment and typically provides higher growth rates than KGM™ Medium.
- Specially designed to support the growth of human and animal primary derived cells.
- KGM-2 Bulletkit™, also sold as separate basal medium and supplements.
Specifications
Red | |
Keratinocyte Cells | |
500 mL Bottle | |
500 mL |
Store in dark at 2 to 8°C | |
Basal Medium | |
7.25 to 7.65 |